missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Indium(III) oxide, 99.9997%, (trace metal basis), Thermo Scientific™
Supplier: Thermo Scientific Chemicals 193170250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
for ultra trace analysisSpecifications
Indium(III) oxide | |
25g | |
In2O3 | |
15,4992 | |
277.63 | |
99.9997% | |
Glass bottle |
1312-43-2 | |
81.0 to 84.3% (In) (Complexometry) | |
In2O3 | |
Solubility in water: insoluble | |
277.63 | |
Trace Metal Basis |
Chemical Identifiers
1312-43-2 | |
In2O3 | |
277.63 |
Safety and Handling
EINECSNumber : 215-193-9
RUO â Research Use Only