missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Indium(III) oxide, 99.999% (metals basis), Thermo Scientific Chemicals
$164.22 - $797.63
Chemical Identifiers
CAS | 1312-43-2 |
---|---|
Molecular Formula | In2O3 |
Molecular Weight (g/mol) | 277.63 |
MDL Number | MFCD00011060 |
InChI Key | PJXISJQVUVHSOJ-UHFFFAOYSA-N |
IUPAC Name | diindium(3+) trioxidandiide |
SMILES | [O--].[O--].[O--].[In+3].[In+3] |
Description
Finds application in semiconductors and can be used as a resistive element in integrated circuits. It is thermally stable and is suitable for glass, optic and ceramic applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1312-43-2 | |
277.63 | |
PJXISJQVUVHSOJ-UHFFFAOYSA-N | |
[O--].[O--].[O--].[In+3].[In+3] |
In2O3 | |
MFCD00011060 | |
diindium(3+) trioxidandiide |
Specifications
850°C (volat.) | |
Sintered Granules | |
In2O3 | |
14,4952 | |
PJXISJQVUVHSOJ-UHFFFAOYSA-N | |
diindium(3+) trioxidandiide | |
277.64 | |
Odorless | |
Indium(III) oxide |
1312-43-2 | |
10 g | |
MFCD00011060 | |
Insoluble in water. | |
[O--].[O--].[O--].[In+3].[In+3] | |
277.63 | |
99.999% | |
7.18 g/mL | |
(metals basis) |
Safety and Handling
EINECSNumber : 215-193-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only