missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Indium(III) oxide, 99.9% (metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1258109

 View more versions of this product

Catalog No. AA1258109



Specifications

Specifications

Indium(III) oxide
99.994% (Metal basis)
MFCD00011060
Insoluble in water and soluble in acids.
-325 Mesh Powder
Odorless
850° volat.
1312-43-2
In2O3
Yellow powder
277.64
7.18
Yellow
10g

Chemical Identifiers

1312-43-2
In2O3
MFCD00011060
Safety and Handling

Safety and Handling

EINECSNumber : 215-193-9

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents