Learn More
Indium(III) nitrate hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01070814
Description
Indium(III) nitrate hydrate acts as an oxidizing agent. It is used in the manufacture of coordinated polymer films which are used in the preparation of indium(III) oxide nanoparticles. It is used in the synthesis of other indium complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Indium(III) nitrate hydrate | |
207398-97-8 | |
Crystalline Aggregates | |
(metals basis) | |
MFCD00149742 | |
indium iii nitrate hydrate, indium nitrate hydrate, nitric acid, indium 3+ salt, hydrate 2:9, indium 3+ ion hydrate trinitrate, indium nitrate hydrate 4.5, indiumnitratehexahydrate, indium nitrate, hydrous, in no3 33h2o, indiumnitratehydratewhitextl, in.3no3.h2o | |
LKRFCKCBYVZXTC-UHFFFAOYSA-N | |
indium(3+) trinitrate | |
3084148 | |
99.999% | |
Hygroscopic |
100°C (in -H2O) | |
White | |
25 g | |
InN3O9 | |
UN1477 | |
Soluble in water,methanol and ethanol. | |
[In+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
300.83 | |
300.83 (Anhydrous) | |
Puratronic™ |
Chemical Identifiers
207398-97-8 | |
300.83 | |
LKRFCKCBYVZXTC-UHFFFAOYSA-N | |
3084148 |
InN3O9 | |
MFCD00149742 | |
indium iii nitrate hydrate, indium nitrate hydrate, nitric acid, indium 3+ salt, hydrate 2:9, indium 3+ ion hydrate trinitrate, indium nitrate hydrate 4.5, indiumnitratehexahydrate, indium nitrate, hydrous, in no3 33h2o, indiumnitratehydratewhitextl, in.3no3.h2o | |
[In+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Safety and Handling
GHS H Statement
H272-H315-H319
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
P210-P220-P221-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
H272-H315-H319
DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.
EINECSNumber : 237-393-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only