Please login to your online account to display your discounted pricing

Alfa Aesar™ Indium(III) n-octoxide, 98%

Chemical Identifiers

Chemical Name or Material Indium(III) n-octoxide
CAS 106461-03-4
Formula Weight 502.48
MDL Number MFCD26967910
UN Number UN1325
Synonym Indium(III) n-octylate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3752303

  SDS
alfa aesar™
H3752303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3752306

  SDS
alfa aesar™
H3752306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 502.48
Color Yellow
Quantity 1g
Sensitivity Moisture Sensitive
UN Number UN1325
Physical Form Solid
Assay 98%
Melting Point ≈150°C (dec.)
Health Hazard 1 Warning
Synonym Indium(III) n-octylate
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
CAS 106461-03-4
DOT Information Transport Hazard Class: 4.1; Packaging Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
Chemical Name or Material Indium(III) n-octoxide
MDL Number MFCD26967910
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.