missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Indium(III) isopropoxide, 99+% (metals basis), 5% w/v isopropanol

Manufacturer:  Alfa Aesar™ 44562AE

 View more versions of this product

Catalog No. AA44562AE



Specifications

Specifications

Indium (III) isopropoxide
12°C (54°F)
82°C
100mL
(metals basis)
In[OCH(CH3)2]3
UN1219
5% w/v isopropanol
OVZUSPADPSOQQN-UHFFFAOYSA-N
indium(3+);propan-2-olate
57346963
Liquid
0.78 g/mL
Moisture sensitive
Colorless
38218-24-5
C9H21InO3
MFCD00210318
2-propanol, indium 3+ salt 9ci
Miscible with alcohols,ketones and esters.
CC(C)[O-].CC(C)[O-].CC(C)[O-].[In+3]
292.082
292.08
≥99%

Chemical Identifiers

38218-24-5
292.082
OVZUSPADPSOQQN-UHFFFAOYSA-N
57346963
CC(C)[O-].CC(C)[O-].CC(C)[O-].[In+3]
C9H21InO3
MFCD00210318
2-propanol, indium 3+ salt 9ci
indium(3+);propan-2-olate
Safety and Handling

Safety and Handling

GHS H Statement
H225-H319-H336
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c

H225-H319-H335-H336

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL

TSCA : No

Recommended Storage : Ambient temperatures

Documents