missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Indium(III) iodide, ultra dry, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 4710804

 View more versions of this product

Catalog No. AA4710804



Description

Description

  • Ampouled under argon
Specifications

Specifications

Indium(III) iodide
13510-35-5
I3In
UN3260
RMUKCGUDVKEQPL-UHFFFAOYSA-K
triiodoindigane
83539
Beads
4.69g/mL
Ampule
Yellow to Brown
2g
Ultra dry
(metals basis)
MFCD00016151
indium triiodide, indium iii iodide, indium iodide ini3, acmc-20akqd, ini3, indium iii iodide, anhydrous, indium iii iodide, anhydrous, powder trace metals basis
[In](I)(I)I
495.531
495.53
99.999%
−10 Mesh
Light Sensitive & Hygroscopic
210°C

Chemical Identifiers

13510-35-5
495.531
RMUKCGUDVKEQPL-UHFFFAOYSA-K
83539
[In](I)(I)I
I3In
MFCD00016151
indium triiodide, indium iii iodide, indium iodide ini3, acmc-20akqd, ini3, indium iii iodide, anhydrous, indium iii iodide, anhydrous, powder trace metals basis
triiodoindigane
Safety and Handling

Safety and Handling

GHS H Statement
H334-H361-H314-H318-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of damaging fertility or the unborn child.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.

GHS P Statement
P260-P285-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
In case of inadequate ventilation wear respiratory protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

TSCA : TSCA

storageNote1 :

  • Ampouled under argon

SDS Documents