Learn More
Indium(III) iodide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04710818
Description
It is used in the preparation vinyl and methyl indates which undergo regioselective reactions with cinnamyl bromide. It is used as an iodide getter in the Cativa Process.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Specifications
Indium(III) iodide | |
Ultra dry | |
Yellow to Brown | |
50 g | |
I3In | |
UN3260 | |
Soluble in water. | |
[In](I)(I)I | |
495.531 | |
495.53 | |
Ampule | |
4.69 g/mL |
210°C | |
13510-35-5 | |
Beads | |
(metals basis) | |
MFCD00016151 | |
indium triiodide, indium iii iodide, indium iodide ini3, acmc-20akqd, ini3, indium iii iodide, anhydrous, indium iii iodide, anhydrous, powder trace metals basis | |
RMUKCGUDVKEQPL-UHFFFAOYSA-K | |
triiodoindigane | |
83539 | |
99.999% | |
Hygroscopic; Light sensitive |
Chemical Identifiers
13510-35-5 | |
495.531 | |
RMUKCGUDVKEQPL-UHFFFAOYSA-K | |
83539 | |
[In](I)(I)I |
I3In | |
MFCD00016151 | |
indium triiodide, indium iii iodide, indium iodide ini3, acmc-20akqd, ini3, indium iii iodide, anhydrous, indium iii iodide, anhydrous, powder trace metals basis | |
triiodoindigane |
Safety and Handling
GHS H Statement
H334-H361-H314-H318-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of damaging fertility or the unborn child.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
P201-P202-P261-P264b-P271-P272-P280-P281-P285-P302+P352-P304+P340-P305+P351+P338-P308+P313-P333+P313-P342+P311-P362-P501c
H315-H317-H319-H334-H335-H360
DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber : 236-839-6
TSCA : Yes
storageNote1 :
- Ampouled under argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only