missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Indium(III) iodide, Red to yellow crystalline, 99.994% (Metals basis), Alfa Aesar™

$89.91 - $812.70

Chemical Identifiers

CAS 13510-35-5
Molecular Formula I3In
Molecular Weight (g/mol) 495.531
MDL Number MFCD00016151
InChI Key RMUKCGUDVKEQPL-UHFFFAOYSA-K
Synonym acmc-20akqd
PubChem CID 83539
IUPAC Name triiodoindigane
SMILES [In](I)(I)I
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1185404 View Documents Alfa Aesar
1185404
2g Each for $89.91
 
AA1185409 View Documents Alfa Aesar
1185409
10g Each for $305.34
 
AA1185418 View Documents Alfa Aesar
1185418
50g Each for $812.70
 
Specifications

Chemical Identifiers

13510-35-5
495.531
RMUKCGUDVKEQPL-UHFFFAOYSA-K
83539
[In](I)(I)I
I3In
MFCD00016151
acmc-20akqd
triiodoindigane

Specifications

13510-35-5
I3In
acmc-20akqd
[In](I)(I)I
495.531
495.53
Red to Yellow
Indium(III) iodide
99.994% (Metals basis)
MFCD00016151
RMUKCGUDVKEQPL-UHFFFAOYSA-K
triiodoindigane
83539
Crystalline
2g
Safety and Handling

Safety and Handling

H334-H317-H361-H314-H318: May cause allergy or asthma symptoms or breathing difficulties if inhaled. May cause an allergic skin reaction. Suspected of damaging fertility or the unborn child. Causes severe skin burns and eye damage. Causes serious eye damage.

EINECSNumber : 236-839-6