Learn More
Indium(III) fluoride, anhydrous, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04011804
Description
Indium(III) fluoride is employed in the synthesis of non-oxide glasses. It promotes chemoselective addition of TMSCN to aldehydes forming the respective cyanohydrins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
>1200°C | |
∼1170°C | |
White | |
Powder | |
MFCD00016149 | |
indium fluoride, indium trifluoride, indium 3+ trifluoride, acmc-1bcyn, indium 3+ ion trifluoride, fluoride, fluoride, in, fluoride | |
171.82 | |
Odorless | |
4.39 g/mL |
Indium(III) fluoride | |
Anhydrous | |
7783-52-0 | |
2 g | |
14,4959 | |
Slightly soluble in water (0;04g/100mL); Freely soluble in dilute acids | |
96% | |
Moisture sensitive |
Chemical Identifiers
7783-52-0 | |
MFCD00016149 | |
indium fluoride, indium trifluoride, indium 3+ trifluoride, acmc-1bcyn, indium 3+ ion trifluoride, fluoride, fluoride, in, fluoride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H332-H314
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 232-005-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only