Please login to your online account to display your discounted pricing

Alfa Aesar™ Indium(III) ethoxide, 99.9% (metals basis)

Chemical Identifiers

Chemical Name or Material Indium (III) ethoxide
CAS 62763-48-8
Molecular Formula C6H15InO3
Linear Formula In(OCH2CH3)3
Formula Weight 250
MDL Number MFCD00799104
UN Number UN3181
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4476902

  SDS
alfa aesar™
4476902
0.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4476904

  SDS
alfa aesar™
4476904
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4476909

  SDS
alfa aesar™
4476909
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder/Chunks
Formula Weight 250
Health Hazard 3 GHS P Statement
P210-P280g-P305+P351+P338-P337+P313
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
CAS 62763-48-8
Assay Percent Notes (metals basis)
UN Number UN3181
Health Hazard 1 Warning
Molecular Formula C6H15InO3
DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
Color Off-white
Sensitivity Moisture Sensitive
MDL Number MFCD00799104
Melting Point 150°C (dec.)
Linear Formula In(OCH2CH3)3
Health Hazard 2 GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 99.9%
Quantity 0.5g
Chemical Name or Material Indium (III) ethoxide
Safety and Handling

Warning

GHS P Statement
P210-P280g-P305+P351+P338-P337+P313
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.