missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Indium(III) ethoxide, 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 4476909

 View more versions of this product

Catalog No. AA4476909



Specifications

Specifications

Indium (III) ethoxide
White
10g
(metals basis)
In(OCH2CH3)3
UN3181
MCXZOLDSEPCWRB-UHFFFAOYSA-N
triethoxyindigane
16701139
Powder/Chunks
Moisture Sensitive
150°C (decomposition)
62763-48-8
C6H15InO3
MFCD00799104
indium ethoxide, indium triethoxide
CCO[In](OCC)OCC
250.001
250
99.9%

Chemical Identifiers

62763-48-8
250.001
MCXZOLDSEPCWRB-UHFFFAOYSA-N
16701139
CCO[In](OCC)OCC
C6H15InO3
MFCD00799104
indium ethoxide, indium triethoxide
triethoxyindigane
Safety and Handling

Safety and Handling

GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P280g-P305+P351+P338-P337+P313
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.

SDS Documents