missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Indium(III) chloride, White to off-white powder, 99.999% (Metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 4434906

 View more versions of this product

Catalog No. AA4434906



Specifications

Specifications

Indium(III) chloride
99.999% (Metals basis)
MFCD00011058
221.18
3.46
800°C
586°C
10025-82-8
InCl3
Reacts with water.
Powder
Hygroscopic
White
5g

Chemical Identifiers

10025-82-8
InCl3
MFCD00011058
Safety and Handling

Safety and Handling

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 233-043-0

RTECSNumber : NL1400000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents