missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Indium(III) chloride hydrate, 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ 1185930

 View more versions of this product

Catalog No. AA1185930



Specifications

Specifications

Indium(III) chloride hydrate
(metals basis)
MFCD00011058
14,4958
KYCHGXYBBUEKJK-UHFFFAOYSA-K
221.18 (Anhydrous)
99.99%
White
143983-91-9
Cl3H2InO
UN3260
Soluble in water
239.18
Crystalline
Hygroscopic
250g

Chemical Identifiers

143983-91-9
239.18
KYCHGXYBBUEKJK-UHFFFAOYSA-K
Cl3H2InO
MFCD00011058
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

TSCA : TSCA

Documents