Please login to your online account to display your discounted pricing

Alfa Aesar™ Indium(III) chloride, anhydrous, 98+%


Quantity 25g

Manufacturer: alfa aesar™  L1875814

 View more versions of this product

Catalog No. AAL1875814

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 221.18
Odor Odorless
Melting Point 586°C
Chemical Name or Material Indium(III) chloride
Name Note anhydrous
Assay ≥98%
Molecular Formula InCl3
MDL Number MFCD00011058
Merck Index 14,4958
Boiling Point 800°C
Quantity 25g
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
RTECS Number NL1400000
TSCA TSCA
CAS 10025-82-8
UN Number UN3260
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Specifications

Chemical Name or Material Indium(III) chloride
CAS 10025-82-8
Name Note anhydrous
Molecular Formula InCl3
Formula Weight 221.18
MDL Number MFCD00011058
UN Number UN3260
Merck Index 14,4958

Applications
Indium(III) chloride is used in famous organic reactions such as Friedel-Crafts acylation and Diels-Alder reaction as a lewis acid catalyst. It is an efficient catalyst for thioacetalization and its chemoselectivity. It catalyzes the enantioselective ring-opening of meso-aziridines with thiols.

Solubility
Soluble in water and tetrahydrofuran. Sparingly soluble in alcohol and insoluble in ether.

Notes
Hygroscopic. Incompatible with strong acids and strong oxidizing agents. The molten state of indium(III) chloride may conduct electricity.

Accessories

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.