Learn More
Indium(III) 2,4-pentanedionate, 98%, Thermo Scientific Chemicals
Used in catalysts
Supplier: Thermo Scientific Chemicals 01379406
Description
Used in catalysts.Indium(III) 2,4-pentanedionate is used as a precursor for the preparation of indium(III)oxide. It acts as a catalyst in the synthesis of oxindole derivatives. It finds application in gas sensors. Further, it is used in the synthesis of indium tin oxide nanocrystals. In addition to this, it is involved in the preparation of thin-film solar cells by atomic layer chemical vapor deposition with hydrogen sulfide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Indium(III) 2,4-pentanedionate | |
186°C (decomposition) | |
MFCD00013494 | |
acetylacetone indium iii salt, indium iii tris 4-oxo-2-pentene-2-olate | |
CMGVNFXMZKDHDG-UHFFFAOYSA-N | |
indium(3+);(Z)-4-oxopent-2-en-2-olate;(E)-4-oxopent-2-en-2-olate | |
101644361 | |
98% |
14405-45-9 | |
C15H21InO6 | |
5 g | |
Soluble in benzene; Insoluble in water. | |
[In+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O | |
412.15 | |
412.15 | |
Powder |
Chemical Identifiers
14405-45-9 | |
412.15 | |
CMGVNFXMZKDHDG-UHFFFAOYSA-N | |
101644361 | |
[In+3].CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O.CC(=O)[CH-]C(C)=O |
C15H21InO6 | |
MFCD00013494 | |
acetylacetone indium iii salt, indium iii tris 4-oxo-2-pentene-2-olate | |
indium(3+);(Z)-4-oxopent-2-en-2-olate;(E)-4-oxopent-2-en-2-olate |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
EINECSNumber : 238-378-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only