Please login to your online account to display your discounted pricing

Alfa Aesar™ Indium(I) iodide, ultra dry, 99.998% (metals basis)

$312.47 - $312.47

Chemical Identifiers

Chemical Name or Material Indium(I) iodide
CAS 13966-94-4
Name Note Ultra dry
Formula Weight 241.72
MDL Number MFCD00135543
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3570006

 
alfa aesar™
3570006
5g Each for $312.47

AA3570003

 
alfa aesar™
3570003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 241.72
Packaging Ampule
Health Hazard 2 GHS H Statement
H334-H302-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Indium(I) iodide
TSCA TSCA
Mesh Size −10 Mesh
Name Note Ultra dry
Physical Form Beads
Assay 99.998%
Sensitivity Moisture & Light Sensitive
Assay Percent Notes (metals basis)
MDL Number MFCD00135543
CAS 13966-94-4
Melting Point 351°C
Quantity 5g
Health Hazard 1 Danger
Storage Note 1
  • Ampouled under argon

  • Ampouled under argon

Safety and Handling

Danger

GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H334-H302-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

  • Ampouled under argon