Please login to your online account to display your discounted pricing

Alfa Aesar™ Indium(I) bromide, ultra dry, 99.998% (metals basis)

$9.55 - $9.55

Chemical Identifiers

Chemical Name or Material Indium(I) bromide
CAS 14280-53-6
Name Note Ultra dry
Formula Weight 194.73
MDL Number MFCD00064630
UN Number UN3260
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3938806

  SDS
alfa aesar™
3938806
5g Each for $9.55

AA3938814

  SDS
alfa aesar™
3938814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 194.73
Packaging Ampule
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 4.96g/mL
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Quantity 5g
MDL Number MFCD00064630
Mesh Size −10 Mesh
Physical Form Beads
Storage Note 1
  • Ampouled under argon
Melting Point 220°C
Sensitivity Moisture Sensitive
UN Number UN3260
Odor Odorless
Boiling Point 662°C
Assay Percent Notes (metals basis)
Chemical Name or Material Indium(I) bromide
CAS 14280-53-6
Name Note Ultra dry
Assay 99.998%
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
Health Hazard 1 Danger

  • Ampouled under argon

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

  • Ampouled under argon