missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Indium foil, 99.9975% (metals basis), Alfa Aesar™, Puratronic™

Form: approx. 1.8g/50x50mm

Manufacturer:  Alfa Aesar 11386FI

 View more versions of this product

Catalog No. AA11386FI



Specifications

Specifications

Indium
7440-74-6
99.9975% (Metals basis)
MFCD00134048
APFVFJFRJDLVQX-UHFFFAOYSA-N
indium
114.818
CHEBI:30430
Silver-White
50 x 50mm
Silvery-white foil
In
indio
[In]
Odorless
5359967
Foil, 0.1mm (0.004 in.) thick

Chemical Identifiers

7440-74-6
114.818
APFVFJFRJDLVQX-UHFFFAOYSA-N
5359967
indium
In
MFCD00134048
indio
CHEBI:30430
[In]
Safety and Handling

Safety and Handling

EINECSNumber : 231-180-0

Documents