missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Indium foil, 99.99% (metals basis), Alfa Aesar™

Form: approx. 4.6g/50x50mm

Manufacturer:  Alfa Aesar 12206HW

 View more versions of this product

Catalog No. AA12206HW



Specifications

Specifications

Indium
7440-74-6
99.99% (Metals basis)
MFCD00134048
APFVFJFRJDLVQX-UHFFFAOYSA-N
indium
114.818
CHEBI:30430
Silver-White
150 x 300mm
Silvery-white foil
In
indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic
[In]
Odorless
5359967
Foil, 0.127mm (0.005 in.) thick

Chemical Identifiers

7440-74-6
114.818
APFVFJFRJDLVQX-UHFFFAOYSA-N
5359967
indium
In
MFCD00134048
indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic
CHEBI:30430
[In]
Safety and Handling

Safety and Handling

EINECSNumber : 231-180-0

Documents