missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Indium foil, 0.5mm (0.02 in.) thick, Puratronic™, 99.9975% (metals basis)

Manufacturer:  Alfa Aesar™ 11384FI

 View more versions of this product

Catalog No. AA11384FI



Specifications

Specifications

Indium foil
50 x 50mm
7440-74-6
In
indio
[In]
Odorless
5359967
Foil
Puratronic™
≈2.3g/25x25mm
0.5mm (0.02 in.) thick
(metals basis)
MFCD00134048
APFVFJFRJDLVQX-UHFFFAOYSA-N
indium
114.818
CHEBI:30430
99.9975%

Chemical Identifiers

7440-74-6
114.818
APFVFJFRJDLVQX-UHFFFAOYSA-N
5359967
indium
In
MFCD00134048
indio
CHEBI:30430
[In]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents