Please login to your online account to display your discounted pricing

Indium foil, 0.127mm (0.005in) thick, 99.99% (metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Indium foil
Name Note 0.127mm (0.005 in.) thick
MDL Number MFCD00134048
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

AA12206FI

 
alfa aesar
12206FI
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form ≈2.3g/50X50mm
Chemical Name or Material Indium foil
TSCA TSCA
Name Note 0.127mm (0.005 in.) thick
Assay 99.99%
Assay Percent Notes (metals basis)
MDL Number MFCD00134048
Odor Odorless