missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Indium Arsenide, 99.9999% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1293209

 View more versions of this product

Catalog No. AA1293209



Specifications

Specifications

Indium arsenide
1303-11-3
AsIn
Gray polycrystalline pieces
RPQDHPTXJYYUPQ-UHFFFAOYSA-N
indiganylidynearsane
91500
1.5 To 9.5mm Polycrystalline Pieces
Gray
Hardly attacked by mineral acids
99.9999% (Metal basis)
MFCD00016144
indium arsenide, indiam arsenide, indium monoarsenide, indium arsenide inas, arsanylidyneindium, dsstox_cid_3825, dsstox_rid_77199, dsstox_gsid_23825, indium arsenide 1g
[As]#[In]
189.74
189.74
Odorless
10g

Chemical Identifiers

1303-11-3
189.74
RPQDHPTXJYYUPQ-UHFFFAOYSA-N
91500
[As]#[In]
AsIn
MFCD00016144
indium arsenide, indiam arsenide, indium monoarsenide, indium arsenide inas, arsanylidyneindium, dsstox_cid_3825, dsstox_rid_77199, dsstox_gsid_23825, indium arsenide 1g
indiganylidynearsane
Safety and Handling

Safety and Handling

H301-H331-H400-H410: Toxic if swallowed. Toxic if inhaled. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

EINECSNumber : 215-115-3

Documents