Learn More
Indium Arsenide, 99.9999% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01293204
Description
Indium arsenide is used for construction of infrared detectors, for the wavelength range of 1-3μm. It is widely used as terahertz radiation source as it is a strong photo-Dember emitter. Indium arsenide is also used for making of diode laser.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Indium arsenide | |
Hardly attacked by mineral acids | |
Gray | |
2 g | |
AsIn | |
indium arsenide, indiam arsenide, indium monoarsenide, indium arsenide inas, arsanylidyneindium, dsstox_cid_3825, dsstox_rid_77199, dsstox_gsid_23825, indium arsenide 1g | |
RPQDHPTXJYYUPQ-UHFFFAOYSA-N | |
indiganylidynearsane | |
91500 | |
Odorless |
936°C | |
1303-11-3 | |
Pieces | |
99.9999% (Metal basis) | |
MFCD00016144 | |
Insoluble in water. | |
[As]#[In] | |
189.74 | |
189.74 | |
Gray polycrystalline pieces |
Chemical Identifiers
1303-11-3 | |
189.74 | |
RPQDHPTXJYYUPQ-UHFFFAOYSA-N | |
91500 | |
[As]#[In] |
AsIn | |
MFCD00016144 | |
indium arsenide, indiam arsenide, indium monoarsenide, indium arsenide inas, arsanylidyneindium, dsstox_cid_3825, dsstox_rid_77199, dsstox_gsid_23825, indium arsenide 1g | |
indiganylidynearsane |
Safety and Handling
P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c
H301+H331-H350
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 215-115-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only