Learn More
Indium antimonide, 99.999% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01462609
Description
Indium antimonide finds use in infrared detectors, including FLIR systems, thermal imaging cameras, infrared astronomy and in infrared homing missile guidance systems, in fast transistors. It is used in thermal image detectors using photo-electromagnetic detectors or photodiodes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Indium antimonide | |
1312-41-0 | |
10 g | |
InSb | |
UN1549 | |
Insoluble in water. | |
[In]#[Sb] | |
236.578 | |
236.57 | |
Odorless |
535°C | |
Pieces | |
(metals basis) | |
MFCD00016146 | |
14,4948 | |
WPYVAWXEWQSOGY-UHFFFAOYSA-N | |
indiganylidynestibane | |
3468413 | |
99.999% | |
5.75 g/mL at m.p. |
Chemical Identifiers
1312-41-0 | |
236.578 | |
WPYVAWXEWQSOGY-UHFFFAOYSA-N | |
indiganylidynestibane |
InSb | |
MFCD00016146 | |
3468413 | |
[In]#[Sb] |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
H302+H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY COMPOUNDS, INORGANIC, SOLID, N.O.S.
EINECSNumber : 215-192-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only