missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Indium, 99.999%, Powder, ACROS Organics™

Manufacturer:  Acros Organics AC193180250

 View more versions of this product

Catalog No. AC193180250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

for ultra trace analysis
Specifications

Specifications

Indium
25g
7440-74-6
100.00%
In
indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic
[In]
114.818
5359967
114.82
99.999%
0.001% max.
156.0°C
7.3
Product contains 2% MgO as anticaking agent
7.3000g/mL
In
06,293; 14,181; 16,181; 17,152
APFVFJFRJDLVQX-UHFFFAOYSA-N
indium
Glass bottle
CHEBI:30430
Powder
Trace Metal Basis
Gray
2000.0°C

Chemical Identifiers

7440-74-6
114.818
indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic
CHEBI:30430
[In]
In
APFVFJFRJDLVQX-UHFFFAOYSA-N
5359967
indium
SDS Documents