missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Indium, 99.999%, Powder, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 193180050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysisSpecifications
Gray | |
156.0°C | |
2000.0°C | |
100.00% | |
In | |
5 g | |
indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic | |
[In] | |
114.82 | |
CHEBI:30430 | |
99.999% | |
Glass bottle |
Indium | |
7440-74-6 | |
7.3000g/mL | |
In | |
MFCD00134048 | |
06,293; 14,181; 16,181; 17,152 | |
APFVFJFRJDLVQX-UHFFFAOYSA-N | |
indium | |
5359967 | |
114.82 | |
Trace Metal Basis | |
Powder |
Chemical Identifiers
7440-74-6 | |
114.82 | |
APFVFJFRJDLVQX-UHFFFAOYSA-N | |
5359967 | |
indium |
In | |
MFCD00134048 | |
indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic | |
CHEBI:30430 | |
[In] |
RUO – Research Use Only