Please login to your online account to display your discounted pricing

Indium, 99.999%, Powder, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC193181000

 View more versions of this product

Catalog No. AC193181000

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Trace Metal Basis
Physical Form Powder
Formula Weight 114.82
Quantity 100g
Color Gray
Density 7.3g/mL
Fieser 06,293; 14,181; 16,181; 17,152
Chemical Name or Material Indium
Molecular Formula In
Linear Formula In
Total Trace Metal Impurities 0.001% max.
Additive Product contains 2% MgO as anticaking agent
CAS 7440-74-6
Packaging Glass bottle
Assay Percent Range 100.00%

Specifications

Chemical Name or Material Indium
CAS 7440-74-6
Purity Grade Trace Metal Basis
Assay Percent Range 100.00%
Molecular Formula In
Linear Formula In
Formula Weight 114.82