Please login to your online account to display your discounted pricing

Alfa Aesar™ Indirubin-3'-oxime, 98%


Quantity 10mg

Manufacturer: alfa aesar™  J67459MA

 View more versions of this product

Catalog No. AAJ67459MA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 277.28
MDL Number MFCD02683594
Storage Note 1 -30° to -10°C
Chemical Name or Material Indirubin-3'-oxime
CAS 160807-49-8
Color Dark red
Physical Form Solid
Synonym Indirubin-3'-monoxime
Assay 98%
Quantity 10mg

Specifications

Chemical Name or Material Indirubin-3'-oxime
CAS 160807-49-8
Formula Weight 277.28
MDL Number MFCD02683594
Synonym Indirubin-3'-monoxime
Accessories

-30° to -10°C