Please login to your online account to display your discounted pricing

Indigo Stock Solution, For Ozone, Certified, LabChem™

$80.94 - $80.94

Chemical Identifiers

Chemical Name or Material Indigo Stock Solution
CAS 7732-18-5
CAS Max % 99.82
CAS 7664-38-2
CAS Max % 0.1
CAS 67627-18-5
CAS Max % 0.08
Name Note For Ozone
Purity Grade Certified Grade
Linear Formula C16H7K3N2O11S3
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC155871

 
lab chem
LC155871
Amber Poly 500mL Each for $80.94
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 616.72
Absorbance Passes Test
Chemical Name or Material Indigo Stock Solution
Quantity 500mL
CAS Max % 0.08
Name Note For Ozone
Physical Form Liquid
Linear Formula C16H7K3N2O11S3
CAS 7732-18-5
Identification Passes Test
Packaging Amber Poly
CAS Max % 99.82
Health Hazard 2 GHS H Statement
Solution is not hazardous.
CAS 7664-38-2
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Solubility Soluble in water
Specific Gravity 1g/mL
CAS Max % 0.1
Recommended Storage Room Temperature
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide; Phosphorus oxides
Density 1g/mL
CAS 67627-18-5
Color Clear blue
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.

Room Temperature