Please login to your online account to display your discounted pricing

Indicator Solution, For Cyanide, Certified, LabChem™

$54.71 - $54.71

Chemical Identifiers

Chemical Name or Material Indicator Solution
CAS 67-64-1
CAS Max % 99.98
CAS 536-17-4
CAS Max % 0.02
Name Note For Cyanide
Purity Grade Certified Grade
Linear Formula C12H12N2OS2
Formula Weight 264.37
UN Number UN1090
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC155857

 
lab chem
LC155857
Amber Glass 125mL Each for $54.71
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 264.37
Chemical Name or Material Indicator Solution
Quantity 125mL
Flash Point −0.18°C
Name Note For Cyanide
Physical Form Liquid
Linear Formula C12H12N2OS2
CAS 67-64-1
UN Number UN1090
Identification Passes Test
Health Hazard 1 Warning
Packaging Amber Glass
CAS Max % 99.98
Health Hazard 2 GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Viscosity 0.417mm2/s
CAS 536-17-4
Health Hazard 3 GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Use only outdoors or in a well-ventilated area.
Avoid breathing vapors.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
In case of fire: Use alcohol resistant foam, extinguishing powder to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
Solubility Soluble in water
Specific Gravity 786kg/m3
Suitability Passes Test
CAS Max % 0.02
Recommended Storage Room Temperature
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide
Density 786kg/m3
Boiling Point 56°C
Melting Point −95°C
Color Clear, yellow to orange
Safety and Handling

Warning

GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, ventilating, lighting equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Use only outdoors or in a well-ventilated area.
Avoid breathing vapors.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
In case of fire: Use alcohol resistant foam, extinguishing powder to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.

Room Temperature