Index of Refraction Plate
An ideal prism to demonstrate the principles of the refractive index.
Manufacturer:  Fisher Science Education 234X234
 View more versions of this product
Catalog No. S42452
Description
Description
High-quality prism, ideal for experiments on the principles of optics and index of refraction. For use by individual students.
  • One set of edges is polished and the opposing set ground
  • Pin-sighting method used to determine index of refraction
  • Plates: 2.8 × 2.8 in. (70 × 70mm), 0.23 or 0.35 in. (6 or 9mm) thick
Specifications
Specifications
Refraction Plate
Each
70 mm
Demonstrate the principles of the refractive index
70 mm