missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Indene, 90%, tech., stabilized, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 122140050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Indene | |
95-13-6,98-29-3 | |
White to Yellow | |
181.0°C | |
Authentic | |
Glass bottle | |
1.5670 to 1.5720 | |
5 g | |
0.99 | |
indene, indonaphthene, inden, unii-67h8y6lb8a, indenyl radical, 1h-inden, pubchem20728, indene, reag, acmc-209ruy | |
YBYIRNPNPLQARY-UHFFFAOYSA-N | |
1H-indene | |
5 to 10ppm tert-butylcatechol | |
CHEBI:41921 | |
90% | |
Crystalline Powder |
tech., 90% | |
-5°C to 3°C | |
0.9900g/mL | |
56°C | |
88% min. (GC) | |
C9H8 | |
MFCD00003777 | |
05, 515 | |
15, 4978 | |
Solubility in water: insoluble. Other solubilities: miscible with most organic solvents,soluble in acetone,miscible with ethanol and ethyl ether | |
C1C=CC2=CC=CC=C21 | |
116.16 | |
7219 | |
116.16 | |
Technical |
Chemical Identifiers
95-13-6 | |
116.16 | |
YBYIRNPNPLQARY-UHFFFAOYSA-N | |
7219 | |
1H-indene |
C9H8 | |
MFCD00003777 | |
indene, indonaphthene, inden, unii-67h8y6lb8a, indenyl radical, 1h-inden, pubchem20728, indene, reag, acmc-209ruy | |
CHEBI:41921 | |
C1C=CC2=CC=CC=C21 |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
GHS Signal Word: Warning
EINECSNumber : 202-393-6
RTECSNumber : NK8225000
TSCA : TSCA
RUO – Research Use Only