missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Indazole, 96%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 122120050

 View more versions of this product

Catalog No. AC122120050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Indazole
Glass bottle
Beige to White
5g
96%
23,122
1,2-benzodiazole, 1,2-benzopyrazole, 1,2-diazaindene, 1h-benzopyrazole, 1h-indazol, 2-azaindole, 2h-indazole, benzopyrazole, indazole, isoindazole
BAXOFTOLAUCFNW-UHFFFAOYSA-N
1H-indazole
9221
118.14
96%
Authentic
270.0°C (743.0 mmHg)
146.0°C to 150.0°C
271-44-3
C7H6N2
15,4976
Solubility in water: soluble in hot water. Other solubilities: soluble in alcohol and ether
C1=CC=C2C(=C1)C=NN2
118.14
CHEBI:36669
Needle-Like Crystalline Powder

Chemical Identifiers

271-44-3
118.14
1,2-benzodiazole, 1,2-benzopyrazole, 1,2-diazaindene, 1h-benzopyrazole, 1h-indazol, 2-azaindole, 2h-indazole, benzopyrazole, indazole, isoindazole
CHEBI:36669
C1=CC=C2C(=C1)C=NN2
C7H6N2
BAXOFTOLAUCFNW-UHFFFAOYSA-N
9221
1H-indazole
Safety and Handling

Safety and Handling

EINECSNumber : 205-978-4

SDS Documents