missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Indazole-3-carboxylic Acid, TCI America™

$27.50 - $80.97

Chemical Identifiers

CAS 4498-67-3
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.148
MDL Number MFCD00211066
InChI Key BHXVYTQDWMQVBI-UHFFFAOYSA-N
Synonym indazole-3-carboxylic acid, 3-carboxyindazole, 3-indazolecarboxylic acid, indazole-3-carboxylicacid, 1h-indazol-3-carbonic acid, 2h-indazole-3-carboxylic acid, 3-carboxy-1h-indazole, 1h-indazole-3-carbonic acid, 1-h-indazole-3-carboxylic acid, benzopyrazole-3-carboxylic acid
PubChem CID 78250
IUPAC Name 1H-indazole-3-carboxylic acid
SMILES C1=CC=C2C(=C1)C(=NN2)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I067225G View Documents TCI America
I067225G
25g Each for $80.97
 
I06725G View Documents TCI America
I06725G
5g Each for $27.50
 
Specifications

Chemical Identifiers

4498-67-3
162.148
BHXVYTQDWMQVBI-UHFFFAOYSA-N
78250
C1=CC=C2C(=C1)C(=NN2)C(=O)O
C8H6N2O2
MFCD00211066
indazole-3-carboxylic acid, 3-carboxyindazole, 3-indazolecarboxylic acid, indazole-3-carboxylicacid, 1h-indazol-3-carbonic acid, 2h-indazole-3-carboxylic acid, 3-carboxy-1h-indazole, 1h-indazole-3-carbonic acid, 1-h-indazole-3-carboxylic acid, benzopyrazole-3-carboxylic acid
1H-indazole-3-carboxylic acid

Specifications

Undesignated
25g
C8H6N2O2
indazole-3-carboxylic acid, 3-carboxyindazole, 3-indazolecarboxylic acid, indazole-3-carboxylicacid, 1h-indazol-3-carbonic acid, 2h-indazole-3-carboxylic acid, 3-carboxy-1h-indazole, 1h-indazole-3-carbonic acid, 1-h-indazole-3-carboxylic acid, benzopyrazole-3-carboxylic acid
C1=CC=C2C(=C1)C(=NN2)C(=O)O
162.148
162.15
≥98.0% (HPLC,T)
261°C
4498-67-3
MFCD00211066
BHXVYTQDWMQVBI-UHFFFAOYSA-N
1H-indazole-3-carboxylic acid
78250
Crystal-Powder at 20°C
Indazole-3-carboxylic Acid
Safety and Handling

Safety and Handling

TSCA : No