Learn More
Indane, 95%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 292480050
Specifications
Indane | |
50°C | |
Glass bottle | |
0.96 | |
Colorless to Yellow | |
5g | |
496-11-7 | |
C9H10 | |
05,II,376 | |
1,2-hydrindene, 1h-indene, 2,3-dihydro, 2,3-dihydroindene, benzocyclopentane, hydrindene, hydrindonaphthene, indan, indane, indane alkane, indene, 2,3-dihydro | |
PQNFLJBBNBOBRQ-UHFFFAOYSA-N | |
2,3-dihydro-1H-indene | |
10326 | |
118.18 | |
95% |
0.9600g/mL | |
Authentic | |
1.5350 to 1.538 | |
176°C | |
-51°C | |
95% | |
94% min. (GC) | |
MFCD00003795 | |
15, 4971 | |
Solubility in water: insoluble. Other solubilities: misciple in ethanol,diethyl ether;,slightly soluble in chloroform | |
C1CC2=CC=CC=C2C1 | |
118.18 | |
CHEBI:37911 | |
Liquid |
Chemical Identifiers
496-11-7 | |
118.18 | |
PQNFLJBBNBOBRQ-UHFFFAOYSA-N | |
10326 | |
2,3-dihydro-1H-indene |
C9H10 | |
MFCD00003795 | |
1,2-hydrindene, 1h-indene, 2,3-dihydro, 2,3-dihydroindene, benzocyclopentane, hydrindene, hydrindonaphthene, indan, indane, indane alkane, indene, 2,3-dihydro | |
CHEBI:37911 | |
C1CC2=CC=CC=C2C1 |
Safety and Handling
GHS H Statement:
Flammable liquid and vapor.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 207-814-7
RTECSNumber : NK3750000
TSCA : TSCA