missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Indane, 95%

$89.05 - $1,043.07

Chemical Identifiers

CAS 496-11-7
Molecular Formula C9H10
Molecular Weight (g/mol) 118.179
MDL Number MFCD00003795
InChI Key PQNFLJBBNBOBRQ-UHFFFAOYSA-N
Synonym 1,2-hydrindene, 1h-indene, 2,3-dihydro, 2,3-dihydroindene, benzocyclopentane, hydrindene, hydrindonaphthene, indan, indane, indane alkane, indene, 2,3-dihydro
PubChem CID 10326
ChEBI CHEBI:37911
IUPAC Name 2,3-dihydro-1H-indene
SMILES C1CC2=CC=CC=C2C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB21442AE
View Documents
Alfa Aesar™
B21442AE
100mL Each for $89.05
 
AAB21442AP
View Documents
Alfa Aesar™
B21442AP
500mL Each for $305.36
 
AAB214420F
View Documents
Alfa Aesar™
B214420F
2500mL Each for $1,043.07
 
Description

Description

Applications
Indane, is used as a catalytic agent, Petrochemical additive, used in organic synthesis. It is also used as pharmaceutical and chemical intermediate.

Solubility
Soluble in ethanol, diethyl ether, chloroform (slightly), and ether. Insoluble in water.

Notes
Keep away from acids, acid chlorides, acid anhydrides, strong oxidizing agents, carbon dioxide. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

496-11-7
118.179
PQNFLJBBNBOBRQ-UHFFFAOYSA-N
10326
2,3-dihydro-1H-indene
C9H10
MFCD00003795
1,2-hydrindene, 1h-indene, 2,3-dihydro, 2,3-dihydroindene, benzocyclopentane, hydrindene, hydrindonaphthene, indan, indane, indane alkane, indene, 2,3-dihydro
CHEBI:37911
C1CC2=CC=CC=C2C1

Specifications

0.962
1.537
-51°C
496-11-7
MFCD00003795
1904376
1,2-hydrindene, 1h-indene, 2,3-dihydro, 2,3-dihydroindene, benzocyclopentane, hydrindene, hydrindonaphthene, indan, indane, indane alkane, indene, 2,3-dihydro
PQNFLJBBNBOBRQ-UHFFFAOYSA-N
2,3-dihydro-1H-indene
10326
118.18
Indane
50°C (122°F)
175°C to 177°C
100mL
C9H10
UN3295
14,4931
Soluble in ethanol,diethyl ether,chloroform (slightly),and ether. Insoluble in water.
C1CC2=CC=CC=C2C1
118.179
CHEBI:37911
95%
Safety and Handling

Safety and Handling

GHS H Statement
H304-H226
May be fatal if swallowed and enters airways.
Flammable liquid and vapour.

P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c

H226-H304

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

EINECSNumber : 207-814-7

RTECSNumber : NK3750000

TSCA : Yes

Recommended Storage : Ambient temperatures