missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Indane, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B21442AE

 View more versions of this product

Catalog No. AAB21442AE



Description

Description

Indane, is used as a catalytic agent, Petrochemical additive, used in organic synthesis. It is also used as pharmaceutical and chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Indane, is used as a catalytic agent, Petrochemical additive, used in organic synthesis. It is also used as pharmaceutical and chemical intermediate.

Solubility
Soluble in ethanol, diethyl ether, chloroform (slightly), and ether. Insoluble in water.

Notes
Keep away from acids, acid chlorides, acid anhydrides, strong oxidizing agents, carbon dioxide. Keep away from strong oxidizing agents.
Specifications

Specifications

Indane
-51°C
175°C to 177°C
C9H10
MFCD00003795
UN3295
14,4931
Soluble in ethanol,diethyl ether,chloroform (slightly),and ether. Insoluble in water.
C1CC2=CC=CC=C2C1
118.179
CHEBI:37911
95%
496-11-7
0.962
50°C (122°F)
1.537
100 mL
1904376
indan, indane, hydrindene, benzocyclopentane, 1h-indene, 2,3-dihydro, 2,3-dihydroindene, 1,2-hydrindene, hydrindonaphthene, indane alkane, indene, 2,3-dihydro
PQNFLJBBNBOBRQ-UHFFFAOYSA-N
2,3-dihydro-1H-indene
10326
118.18

Chemical Identifiers

496-11-7
118.179
PQNFLJBBNBOBRQ-UHFFFAOYSA-N
10326
2,3-dihydro-1H-indene
C9H10
MFCD00003795
indan, indane, hydrindene, benzocyclopentane, 1h-indene, 2,3-dihydro, 2,3-dihydroindene, 1,2-hydrindene, hydrindonaphthene, indane alkane, indene, 2,3-dihydro
CHEBI:37911
C1CC2=CC=CC=C2C1
Safety and Handling

Safety and Handling

GHS H Statement
H304-H226
May be fatal if swallowed and enters airways.
Flammable liquid and vapour.

P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c

H226-H304

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

EINECSNumber : 207-814-7

RTECSNumber : NK3750000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only