missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Indane-5-sulfonyl chloride, 97%, ACROS Organics™

Manufacturer:  Acros Organics AC437990010

 View more versions of this product

Catalog No. AC437990010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Indane-5-sulfonyl chloride
Glass bottle
47.8°C to 48.8°C
52205-85-3
C9H9ClO2S
SWLIXMXSCZYVTQ-UHFFFAOYSA-N
2,3-dihydro-1H-indene-5-sulfonyl chloride
3142583
Crystalline Powder
Authentic
White
1g
97%
indane-5-sulfonyl chloride, indan-5-sulfonyl chloride, indan-5-sulfonylchloride, 5-indanesulfonoyl chloride, chloroindan-5-ylsulfone, 1h-indene-5-sulfonyl chloride, 2,3-dihydro, indanesulfonoylchloride, pubchem5538, 5-chlorosulphonyl indane, acmc-1av5j
C1CC2=C(C1)C=C(C=C2)S(=O)(=O)Cl
216.679
216.69
97%

Chemical Identifiers

52205-85-3
216.679
indane-5-sulfonyl chloride, indan-5-sulfonyl chloride, indan-5-sulfonylchloride, 5-indanesulfonoyl chloride, chloroindan-5-ylsulfone, 1h-indene-5-sulfonyl chloride, 2,3-dihydro, indanesulfonoylchloride, pubchem5538, 5-chlorosulphonyl indane, acmc-1av5j
2,3-dihydro-1H-indene-5-sulfonyl chloride
C9H9ClO2S
SWLIXMXSCZYVTQ-UHFFFAOYSA-N
3142583
C1CC2=C(C1)C=C(C=C2)S(=O)(=O)Cl
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

Documents