missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Indacaterol Maleate 98.0+%, TCI America™

$712.66 - $712.66

Chemical Identifiers

CAS 753498-25-8
Molecular Formula C28H32N2O7
Molecular Weight (g/mol) 508.571
MDL Number MFCD20526769
InChI Key IREJFXIHXRZFER-FTBISJDPSA-N
Synonym (R)-5-[2-[(5,6-Diethyl-2,3-dihydro-1H-inden-2-yl)amino]-1-hydroxyethyl]-8-hydroxyquinolin-2(1H)-one Maleate
PubChem CID 66713197
IUPAC Name but-2-enedioic acid;5-[(1R)-2-[(5,6-diethyl-2,3-dihydro-1H-inden-2-yl)amino]-1-hydroxyethyl]-8-hydroxy-1H-quinolin-2-one
SMILES CCC1=C(C=C2CC(CC2=C1)NCC(C3=C4C=CC(=O)NC4=C(C=C3)O)O)CC.C(=CC(=O)O)C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I0927100MG View Documents Tci America
I0927100MG
100mg Each for $712.66
 
Specifications

Chemical Identifiers

753498-25-8
508.571
IREJFXIHXRZFER-FTBISJDPSA-N
66713197
CCC1=C(C=C2CC(CC2=C1)NCC(C3=C4C=CC(=O)NC4=C(C=C3)O)O)CC.C(=CC(=O)O)C(=O)O
C28H32N2O7
MFCD20526769
(R)-5-[2-[(5,6-Diethyl-2,3-dihydro-1H-inden-2-yl)amino]-1-hydroxyethyl]-8-hydroxyquinolin-2(1H)-one Maleate
but-2-enedioic acid;5-[(1R)-2-[(5,6-diethyl-2,3-dihydro-1H-inden-2-yl)amino]-1-hydroxyethyl]-8-hydroxy-1H-quinolin-2-one

Specifications

White-Yellow
100mg
C28H32N2O7
(R)-5-[2-[(5,6-Diethyl-2,3-dihydro-1H-inden-2-yl)amino]-1-hydroxyethyl]-8-hydroxyquinolin-2(1H)-one Maleate
CCC1=C(C=C2CC(CC2=C1)NCC(C3=C4C=CC(=O)NC4=C(C=C3)O)O)CC.C(=CC(=O)O)C(=O)O
508.571
508.57
≥98.0% (HPLC)
186°C
753498-25-8
MFCD20526769
IREJFXIHXRZFER-FTBISJDPSA-N
but-2-enedioic acid;5-[(1R)-2-[(5,6-diethyl-2,3-dihydro-1H-inden-2-yl)amino]-1-hydroxyethyl]-8-hydroxy-1H-quinolin-2-one
66713197
Crystal-Powder at 20°C
Indacaterol Maleate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator