Please login to your online account to display your discounted pricing

INCB 3284 dimesylate, Tocris™

Selective CCR2 antagonist

$284.81 - $1,180.32

Chemical Identifiers

Chemical Name or Material INCB 3284 dimesylate
Assay Percent Range >98%
Molecular Formula C26H31F3N4O4·2CH4O3S
Formula Weight 712.76
Synonym N-[2-[[(3R)-1-[trans-4-Hydroxy-4-(6-methoxy-3-pyridinyl)cyclohexyl]-3-pyrrolidinyl]amino]-2-oxoethyl]-3-(trifluoromethyl)-benzamide dimethanesulfonate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

430610

 
r & d systems
RDS430610
10mg Each for $284.81

430650

 
r & d systems
RDS430650
50mg Each for $1,180.32
Description & Specifications

Specifications

Formula Weight 712.76
Quantity 10mg
CAS 887401-93-6
Molecular Formula C26H31F3N4O4·2CH4O3S
Assay Percent Range >98%
Synonym N-[2-[[(3R)-1-[trans-4-Hydroxy-4-(6-methoxy-3-pyridinyl)cyclohexyl]-3-pyrrolidinyl]amino]-2-oxoethyl]-3-(trifluoromethyl)-benzamide dimethanesulfonate
Chemical Name or Material INCB 3284 dimesylate
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C