Please login to your online account to display your discounted pricing

Impentamine dihydrobromide, Tocris

Selective H3 antagonist

$177.37 - $746.48

Chemical Identifiers

Chemical Name or Material Impentamine dihydrobromide
Assay Percent Range >98%
Molecular Formula C8H15N3·2HBr
Formula Weight 315.05
Synonym 4-(5-Aminopentyl)imidazole dihydrobromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

185810

 
r & d systems
RDS185810
10mg Each for $177.37

185850

 
r & d systems
RDS185850
50mg Each for $746.48
Description & Specifications

Specifications

Formula Weight 315.05
Quantity 10mg
Chemical Name or Material Impentamine dihydrobromide
Assay Percent Range >98%
Synonym 4-(5-Aminopentyl)imidazole dihydrobromide
Molecular Formula C8H15N3·2HBr
CAS 149629-70-9
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature