Impacto™ Wrist support with fingerless glove
Impacto™ 705-20 Fingerless Nylon glove with adjustable wrist support to prevent from RSI
Manufacturer:  Impacto™ 70520RMED
 View more versions of this product
Catalog No. 19-062980
Description
Description
Impacto™ Wrist Support
  • Fingerless nylon Lycra glove with grain leather palm
  • Grain leather wrist support prevents wrist over flexion and over extension to protect from RSI
Specifications
Specifications
Right
5 in.
Impacto wrist support
Leather
12.7 cm
Medium