Please login to your online account to display your discounted pricing
Impacto™ Half-Finger Impact Gloves
With impact absorbing visco-elastic polymer padding in the palm
$27.61 - $27.61
Specifications
Material Leather
Material Nylon Lycra™
Exterior Finish Soft
Finger Style Half
Includes Visco Elastic Polymer padding in the palm
View More Specs
Products
Catalog Number Mfr. No. Circumference (English) Hand Length (English) Size Price Quantity    
19-031822
impacto protective
40130RMED
8 to 9 in. 8 in. Medium Each for $27.61
Description & Specifications
Specifications
Material Leather
Material Nylon Lycra™
Exterior Finish Soft
Finger Style Half
Includes Visco Elastic Polymer padding in the palm
  • Half finger nylon Lycra and pearl leather glove
  • Adjustable hook and loop wrist closure
  • Provides protection from impact and shock
  • Used with hand tools