Impacto™ Back Coach Back Support
Impacto™ Back Coach Back support with suspenders support the lower back and abdominal area
Manufacturer: Impacto™ BCMED
Description
Impacto™ Back Coach Back Support
- Features adjustable elastic side straps
- Non-slip strips, nonmetallic support stays offer additional support
- Mesh panels for ventilation
- Suspenders included
Order Info
When ordering belts to be worn over clothing, add 2 in. (5.1cm) to waist size.
Specifications
Hook and loop | |
Elastic | |
81.3 to 94 cm | |
Medium |
Breakaway suspenders, light-duty applications, used in many industries including retail, warehousing, manufacturing, retail, automotive, provides protection from strain when working on repetitive tasks or lifting | |
32 to 37 in. | |
Back Coach™ Back Support |