missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Impacto™ Airsol Anti-Fatigue Molded Insoles

Impacto™ AIRSOL Antifatigue Insoles provides relief from fatigue for long term walking or standing on cold, hard surfaces

Manufacturer:  Impacto™ ASMOLDA

 View more versions of this product

Catalog No. 19-088-1238A



Description

Description

Impacto™ Airsol Molded Antifatigue insoles

  • Open-cell structure for better breathability and reducing heat build-up
  • Top layer cushioning provides even distribution of weight to protect pressure points
  • Provide superior impact and shock absorption
  • Used with safety footwear, rubber boots and running shoes
Specifications

Specifications

Women's
Green
Safety footwear, boots and running shoes
4.5 in.
Foam
Molded
38.1 cm
Impacto Insoles
4.5 x 4 in. (11.43 x 38.1 cm)
Industrial
11.43 cm
5 to 6.5
4 in.
0.5 in.
SDS
Documents
Product Certifications