missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Impacto™ Airsol Anti-Fatigue Flat Insoles

Impacto™ AIRSOL ASFLAT Anti-fatigue insoles provide full foot comfort and cushioning with low cut and dress shoes

Manufacturer:  Impacto™ ASFLATB

 View more versions of this product

Catalog No. 19-088-1237B



Description

Description

Impacto™ Airsol Anti-Fatigue Insoles

  • Cellular cushioning open cell foam
  • Biocide additive helps prevent foot odor
  • Full flat design for use with dress and low cut footwear
  • Helps relieve foot, legs and back fatigue
  • Great for daily wear with low cut and dress shoes
Specifications

Specifications

Women's
Green
For relieve foot, legs and back fatigue
Industrial
16.51 cm
7 to 8.5
4 in.
0.25 in.
Insoles
6.5 x 4 in. (16.51 x 10.6 cm)
Dress and low cut footwear
6.5 in.
Open-cell foam
Flat
10.6 cm
SDS
Documents
Product Certifications