missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Impacto™ Airsol Anti-Fatigue Flat Insoles

Impacto™ AIRSOL ASFLAT Anti-fatigue insoles provide full foot comfort and cushioning with low cut and dress shoes

Manufacturer:  Impacto™ ASFLATG

 View more versions of this product

Catalog No. 19-088-1235C



Description

Description

Impacto™ Airsol Anti-Fatigue Insoles

  • Cellular cushioning open cell foam
  • Biocide additive helps prevent foot odor
  • Full flat design for use with dress and low cut footwear
  • Helps relieve foot, legs and back fatigue
  • Great for daily wear with low cut and dress shoes
Specifications

Specifications

Men's
12.5 x 3 in. (31.75 x 10.16 cm)
Dress and low cut footwear
12.5 in.
Open-cell foam
Flat
10.16 cm
Green
For relieve foot, legs and back fatigue
Industrial
31.75 cm
11 to 12.5
3 in.
0.25 in.
SDS
Documents