Please login to your online account to display your discounted pricing

Thermo Scientific™ Immuno Clear Standard Modules


Perform applications which are delivering well-to-well reproducibility with Immuno Clear Standard Modules. Manufactured to fit standard equipment.

Manufacturer: thermo scientific™  N475078

 View more versions of this product

 Disclaimers

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Catalog No. 12-565-117

  • / Case of 60 Each
 

Description & Specifications

Specifications

Certifications/Compliance Certified for adsorption of IgG; CV of <5% between wells
Surface Coating MaxiSorp
Surface Coating Polystyrene
Binding Property Adsorption
Binding Property Hydrophilic
Plate Blocking None
No. per Case 60
No. per Pack 5
Detection Method Colorimetric
Format 96-well strip plate
Color Clear
Well Design U8
Type Framed
Certified Yes
Volume (Metric) Well 300µL
Volume (Metric) Working 250µL

Perform applications which are delivering well-to-well reproducibility with Thermo Scientific™ Immuno Clear Standard Modules, which feature Nunc™, Immulon™ and Microtiter™ module plates developed for ELISA techniques. Manufactured to fit standard equipment, modules provide flexible use of components with small sample sizes. They are backed by industry-leading quality standards and certifications.

Thermo Scientific standard modules are strip plates with solid modules that can be easily inserted and removed from their frame. This enhances the use of the plate with smaller sample sizes.

  • Several strip formats: 8-well strip, 12-well strip, 16-well strip (two columns of 8 wells)
  • Several well shapes: flat bottom, C bottom (radius edge), U shape wells, StarWells (wells have fins to increase the surface area and enhance performance)
  • Several surfaces to maximize assay performance
  • Modules remain in frame when inverted
  • Stackable frame marked for lid orientation
  • Polystyrene wells in polyethylene frame
  • Ext. Dim.: 128 × 86mm

Choice of Surface

  • PolySorp™, Immulon 1B, and Universal Binding (UB) for adsorption of hydrophobic molecules
  • Medisorp™ and Immulon 2HB are slightly hydrophilic and bind a diverse range of biomolecules
  • MaxiSorp™, Immulon 4 HBX, and Enhanced Binding(EB) are hydrophilic and ideal for antibody sandwich assays
  • MultiSorp™ are very hydrophilic and works well for glycans and water soluble proteins

Ordering Information:

Free-standing modules. Cat. No. 95029440 and 464394 are certified. Frames and modules available separately. Sealing tape and strip caps are available. Color-coded modules are available on request. Contact your Customer Service or Sales Representative for further details.