MilliporeSigma™ Immobilon Transfer Membranes
MilliporeSigma™ Immobilon Transfer Membranes offer the convenience of pre-cut sheets with the flexibility of membrane rolls
$374.00 - $561.00
Specifications
Item Description Western Blotting Equipment
View More Specs
Products 5
Catalog Number Mfr. No. Description For Use With (Application) Pore Size Price Quantity  
Catalog Number Mfr. No. Description For Use With (Application) Pore Size Price Quantity  
IEVH85R View Documents MilliporeSigma™
IEVH85R
Immobilon-E transfer membrane Chemiluminescence, Chromogenic detection. 0.45μm Each for $506.00
IMDISP View Documents MilliporeSigma™
IMDISP
Membrane dispenser Immunoblotting, Dot blotting N/A Each for $374.00
IPFL85R View Documents MilliporeSigma™
IPFL85R
Immobilon-FL transfer membrane Fluorescence, Chemifluorescence, Chromogenic, Chemiluminescence 0.45μm Each for $561.00
IPVH85R View Documents MilliporeSigma™
IPVH85R
Immobilon-P transfer membrane Chemiluminescence, Chromogenic, Radioactive Detection 0.45μm Each for $506.00
ISEQ85R View Documents MilliporeSigma™
ISEQ85R
Immobilon-PSQ transfer membrane Chemiluminescence, Chromogenic, Radioactive Detection 0.2μm Each for $506.00
Description
Description
  • Single-cut convenience
  • Compatibility with mini or midi gels
  • No waste
  • Compact package saves valuable lab space
Specifications
Specifications
Western Blotting Equipment